Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salicylaldoxime 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20740-0250 20740-1000 20740-5000
TS20740-0250EA 44 USD InStock
TS20740-0250 TS20740-1000 TS20740-5000
Salicylaldoxime 98%
Salicylaldoxime


Formula: C₇H₇NO₂
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00002120
CAS Number: 94-67-7
EINECS: 202-353-8
Order Now
Specification Test Results
Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 54 to 59 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty