Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-ethyl-1H-pyrazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44671-0010 44671-0050
TS44671-0010EA 162 USD InStock
TS44671-0010 TS44671-0050
3-Amino-5-ethyl-1H-pyrazole 97%
3-Amino-5-ethyl-1H-pyrazole


Formula: C₅H₉N₃
Storage Temperature: Freezer
MDL Number: MFCD08060982
CAS Number: 1904-24-1
Order Now
Specification Test Results
Appearance Yellow to orange to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty