Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-1,2,4-thiadiazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

37603-0010 37603-0050 37603-0250
TS37603-0010EA 58 USD InStock
TS37603-0010 TS37603-0050 TS37603-0250
3,5-Dichloro-1,2,4-thiadiazole 97%
3,5-Dichloro-1,2,4-thiadiazole


Formula: C₂Cl₂N₂S
Boiling Pt: 165…167 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00799286
CAS Number: 2254-88-8
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5570 to 1.5600 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty