Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoisobutyryl bromide 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40309-1000 40309-5000 40309-0050
TS40309-1000EA 63 USD InStock
TS40309-1000 TS40309-5000 TS40309-0050
2-Bromoisobutyryl bromide 98%
2-Bromoisobutyryl bromide


Formula: C₄H₆Br₂O
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000122
CAS Number: 20769-85-1
EINECS: 244-017-3
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5100 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty