Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentanesulphonyl chloride 90%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43783-0010 43783-0050
TS43783-0010EA 170 USD InStock
TS43783-0010 TS43783-0050
Cyclopentanesulphonyl chloride 90%
Cyclopentanesulphonyl chloride


Formula: C₅H₉ClO₂S
MW: 168.64 g/mol
Boiling Pt: 95…98 °C (2Torr)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04117774
CAS Number: 26394-17-2
Order Now
Specification Test Results
Appearance Light brown to dark brown Liquid
Infrared spectrum Conforms
GC ≥88.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty