Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nicotinoyl chloride hydrochloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

33841-0050 33841-0250 33841-1000
TS33841-0050EA 35 USD InStock
TS33841-0050 TS33841-0250 TS33841-1000
Nicotinoyl chloride hydrochloride 97%
Nicotinoyl chloride hydrochloride


Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Melting Pt: 151…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00012820
CAS Number: 20260-53-1
EINECS: 243-648-1
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 156 °C
Titration RCOCl 96.0 to 104.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty