Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloroquinazoline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

46073-0050 46073-0250
TS46073-0050EA 52 USD InStock
TS46073-0050 TS46073-0250
2,4-Dichloroquinazoline 98%
2,4-Dichloroquinazoline


Formula: C₈H₄Cl₂N₂
Melting Pt: 115…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00091950
CAS Number: 607-68-1
Order Now
Specification Test Results
Appearance White to yellow to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 116 to 123 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty