Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropanesulphonyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43902-0010 43902-0050
TS43902-0010EA 345 USD InStock
TS43902-0010 TS43902-0050
Cyclopropanesulphonyl chloride 97%
Cyclopropanesulphonyl chloride


Formula: C₃H₅ClO₂S
MW: 140.59 g/mol
Boiling Pt: 60 °C (2Torr)
Density: 1.38 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01631933
CAS Number: 139631-62-2
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty