Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1H-pyrazole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39682-0010 39682-0050 39682-0250
TS39682-0010EA 54 USD InStock
TS39682-0010 TS39682-0050 TS39682-0250
4-Bromo-1H-pyrazole 98%
4-Bromo-1H-pyrazole


Formula: C₃H₃BrN₂
Boiling Pt: 250…260 °C (760 mmHg)
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00075602
CAS Number: 2075-45-8
Order Now
Specification Test Results
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 99 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty