Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylbutyryl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40982-0050 40982-0250 40982-1000
TS40982-0050EA 28 USD InStock
TS40982-0050 TS40982-0250 TS40982-1000
2-Ethylbutyryl chloride 99%
2-Ethylbutyryl chloride


Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00018812
CAS Number: 2736-40-5
EINECS: 220-359-9
UN: 2920
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
Titration RCOCl ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty