Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxocyclopentanecarboxylic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42634-0010 42634-0050
TS42634-0010EA 176 USD InStock
TS42634-0010 TS42634-0050
3-Oxocyclopentanecarboxylic acid 97%
3-Oxocyclopentanecarboxylic acid


Formula: C6H8O3
Melting Pt: 59…63.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01320173
CAS Number: 98-78-2
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 59 to 64 °C
NMR (1H-NMR) Consistent with structure
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty