Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Methylmorpholine hydrochloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45678-0010 45678-0050
TS45678-0010EA 56 USD InStock
TS45678-0010 TS45678-0050
(R)-3-Methylmorpholine hydrochloride 97%
(R)-3-Methylmorpholine hydrochloride


Formula: C₅H₁₂ClNO
Melting Pt: 126…128.5 °C
Storage Temperature: Ambient
MDL Number: MFCD18382515
CAS Number: 953780-78-4
Order Now
Specification Test Results
Appearance White to yellow to light brown Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
Specific optical rotation +1° to +3° (20°C, 589 nm) c=1 in H2O

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty