Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylthiocarbamoyl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40861-0250 40861-1000
TS40861-0250EA 90 USD InStock
TS40861-0250 TS40861-1000
Dimethylthiocarbamoyl chloride 99%
Dimethylthiocarbamoyl chloride


Formula: C₃H₆ClNS
MW: 123.60 g/mol
Boiling Pt: 63…65 °C (.2mbar)
Melting Pt: 42…43 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00004919
CAS Number: 16420-13-6
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance Yellow Crystals or low melting crystalline solid
Infrared spectrum Conforms
GC ≥98.5 %
Appearance of solution (10% in CHCl3) clear faint yellow to yellow

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty