Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21627-0010 21627-0051 21627-2500
TS21627-0010EA 159 USD InStock
TS21627-0010 TS21627-0051 TS21627-2500
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers) 95%
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)


Formula: C₁₀H₁₀O₃
Boiling Pt: 275 °C (760 mmHg)
Density: 1.23 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00167036
CAS Number: 25134-21-8
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5080 (20°C, 589 nm)
GC ≥94 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty