Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propionyl bromide 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39236-0050 39236-0250 39236-1000
TS39236-0050EA 38 USD InStock
TS39236-0050 TS39236-0250 TS39236-1000
Propionyl bromide 95%
Propionyl bromide


Formula: C₃H₅BrO
Boiling Pt: 104 °C (760 mmHg)
Density: 1.52 g/cm³
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00000116
CAS Number: 598-22-1
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4525 to 1.4575 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty