Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

S-Methyl methanethiosulphonate (MMTS) 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

19739-0010 19739-0050 19739-0250 19739-1000
TS19739-0010EA 43 USD InStock
TS19739-0010 TS19739-0050 TS19739-0250 TS19739-1000
S-Methyl methanethiosulphonate (MMTS) 97%
S-Methyl methanethiosulfonate (MMTS)


Formula: C₂H₆O₂S₂
Boiling Pt: 69…71 °C (.4 mmHg)
Density: 1.227 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007565
CAS Number: 2949-92-0
UN: 3334
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty