Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36781-0010 36781-0050 36781-0250
TS36781-0010EA 36 USD InStock
TS36781-0010 TS36781-0050 TS36781-0250
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 97%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol


Formula: C₉H₇F₆NO
Melting Pt: 147…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039258
CAS Number: 722-92-9
Order Now
Specification Test Results
Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty