Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methyl-3,5-heptadien-2-one 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45321-0050 45321-0250 45321-1000
TS45321-0050EA 27 USD InStock
TS45321-0050 TS45321-0250 TS45321-1000
6-Methyl-3,5-heptadien-2-one 96%
6-Methyl-3,5-heptadien-2-one


Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 168 °C
Density: 0.893 g/cm³
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00043647
CAS Number: 1604-28-0
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5360 (20°C, 589 nm)
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty