Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Methylcarbazole-3-boronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44144-0010 44144-0050
TS44144-0010EA 93 USD InStock
TS44144-0010 TS44144-0050
9-Methylcarbazole-3-boronic acid pinacol ester 97%
9-Methylcarbazole-3-boronic acid pinacol ester


Formula: C₁₉H₂₂BNO₂
Melting Pt: 151.7…152.4 °C
Storage Temperature: Ambient
MDL Number: MFCD16294552
CAS Number: 1217891-71-8
Order Now
Specification Test Results
Appearance Light brown Powder
Infrared spectrum Conforms
Melting point 147 to 154 °C
HPLC ≥96.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty