Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-Catechin monohydrate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46302-0010 46302-0050
TS46302-0010EA 60 USD InStock
TS46302-0010 TS46302-0050
(+)-Catechin monohydrate
(+)-Catechin monohydrate


Formula: C₁₅H₁₄O₆·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00075649
CAS Number: 225937-10-0
Order Now
Specification Test Results
Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤8 %
Solubility (5 % in Ethanol, hot) Clear to hazy
HPLC ≥96.0 %
Specific optical rotation +24° to +28° (20°C, 589 nm) (1 % in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty