Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-methylbenzaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43669-0010 43669-0050
TS43669-0010EA 54 USD InStock
TS43669-0010 TS43669-0050
3-Chloro-4-methylbenzaldehyde 97%
3-Chloro-4-methylbenzaldehyde


Formula: C₈H₇ClO
MW: 154.593 g/mol
Boiling Pt: 232 °C (760 mmHg)
Melting Pt: 37…31 °C
Storage Temperature: Ambient
MDL Number: MFCD06411284
CAS Number: 3411-03-8
Order Now
Specification Test Results
Appearance Colorless to white Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty