Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydropyran-2-ylmethylamine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45337-0010 45337-0050
TS45337-0010EA 190 USD InStock
TS45337-0010 TS45337-0050
Tetrahydropyran-2-ylmethylamine 97%
Tetrahydropyran-2-ylmethylamine


Formula: C₆H₁₃NO
Boiling Pt: 110 °C (25 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06738971
CAS Number: 6628-83-7
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty