Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Amino-7-hydroxytetralin 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43559-0010 43559-0050
TS43559-0010EA 184 USD InStock
TS43559-0010 TS43559-0050
(S)-2-Amino-7-hydroxytetralin 95%
(S)-2-Amino-7-hydroxytetralin


Formula: C₁₀H₁₃NO
Melting Pt: 146.7…150.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11506007
CAS Number: 85951-60-6
Order Now
Specification Test Results
Appearance White to grey Crystalline Powder or crystals
Infrared spectrum Conforms
Melting point 144 to 151 °C
Specific optical rotation -85° to -95° (20 °C, 589 nm) (c=0.5, MeOH)
Titration with HClO4 94 to 106 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty