Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium malonate hydrate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

41350-0250 41350-1000 41350-5000
TS41350-0250EA 50 USD InStock
TS41350-0250 TS41350-1000 TS41350-5000
Disodium malonate hydrate 99%
Disodium malonate hydrate


Formula: C₃H₂Na₂O₄·xH₂O
Storage Temperature: Ambient
CAS Number: 132499-82-2
EINECS: 205-514-0
Order Now
Specification Test Results
Appearance White Crystalline powder and/or chunks
Infrared spectrum Conforms
Water 7 to 13 % (K.F.)
Titration with HClO4 ≥98.5 % (On dry substance)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty