Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzimidazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45167-0050 45167-0250
TS45167-0050EA 127 USD InStock
TS45167-0050 TS45167-0250
5-Bromobenzimidazole 97%
5-Bromobenzimidazole


Formula: C₇H₅BrN₂
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00160001
CAS Number: 4887-88-1
Order Now
Specification Test Results
Appearance Off-white to orange to brown Solid
Infrared spectrum Conforms
Melting point 129 to 132 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty