Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester 90%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44554-0010 44554-0050
TS44554-0010EA 332 USD InStock
TS44554-0010 TS44554-0050
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester 90%
trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronic acid pinacol ester


Formula: C₁₄H₂₅BO₄
Boiling Pt: 79 °C (.1Torr)
Storage Temperature: Ambient
MDL Number: MFCD03788736
CAS Number: 642066-70-4
Order Now
Specification Test Results
Appearance Colorless Oily liquid
GC ≥88.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty