Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycyrrhizic acid monoammonium salt 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

21375-0100 21375-0250 21375-1000
TS21375-0100EA 46 USD InStock
TS21375-0100 TS21375-0250 TS21375-1000
Glycyrrhizic acid monoammonium salt 98+%
Glycyrrhizic acid monoammonium salt


Formula: C₄₂H₆₅NO₁₆
Melting Pt: 209 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00167400
CAS Number: 53956-04-0
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤6.0 % (K.F.)
Heavy metals ≤20 ppm
Sulfated ash ≤0.2 %
Related substances ≤10.0 % 18-alpha-glycyrrhizic acid (HPLC)
Specific optical rotation +49.0° to +54.0° (20°C, 589 nm) (c=1, EtOH 50% v/v) (on anhydrous substance)
Titration with HClO4 98.0 to 102.0 % (on anhydrous substance)

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty