Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzoin 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

10551-0025 10551-1000 10551-5000
TS10551-0025EA 328 USD InStock
TS10551-0025 TS10551-1000 TS10551-5000
Benzoin 98%
Benzoin


Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 194 °C (12 mmHg)
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3
Order Now
Specification Test Results
Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Melting point 133 to 137 °C
Loss on drying ≤1 %
Sulfated ash ≤0.2 %
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty