Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromoethanol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10692-0050 10692-1000 10692-2500
TS10692-0050EA 77 USD InStock
TS10692-0050 TS10692-1000 TS10692-2500
2-Bromoethanol 97%
2-Bromoethanol


Formula: BrH₂CCH₂OH
Boiling Pt: 149…150 °C (750 mmHg)
Melting Pt: –80 °C
Density: 1.76 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002827
CAS Number: 540-51-2
EINECS: 208-748-1
UN: 2927
ADR: 6.1,II
Merck Index: 13,03826
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4940 (20°C, 589 nm)
GC ≥98.5 %
Acid value ≤0.1 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty