Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloropropionyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10999-1000 10999-5000
TS10999-1000EA 59 USD InStock
TS10999-1000 TS10999-5000
3-Chloropropionyl chloride 98%
3-Chloropropionyl chloride


Formula: C₃H₄Cl₂O
MW: 126.97
Boiling Pt: 143
Melting Pt: -32
Density: 1.33 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00000747
CAS Number: 625-36-5
EINECS: 210-890-4
UN: 3489
ADR: 6.1 I
Order Now
Specification Test Results
Appearance Clear colorless to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4590 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer About 0.05% 3,5-di-tert-Butylcatechol

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty