Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutanecarbonyl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11091-0010 11091-0100 11091-0500
TS11091-0010EA 28 USD InStock
TS11091-0010 TS11091-0100 TS11091-0500
Cyclobutanecarbonyl chloride 99%
Cyclobutanecarbonyl chloride


Formula: C₅H₇ClO
Boiling Pt: 145 °C (760 mmHg)
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001319
CAS Number: 5006-22-4
EINECS: 225-680-8
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4545 to 1.4565 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
Absorption Ratio P-15/P+15 1.06 to 1.19
BSC Certification Passes

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty