Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyclopenten-1-one 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11157-0050 11157-0250 11157-1000
TS11157-0050EA 79 USD InStock
TS11157-0050 TS11157-0250 TS11157-1000
2-Cyclopenten-1-one 98%
2-Cyclopenten-1-one


Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 64
Density: 0.98 g/cm³
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001401
CAS Number: 930-30-3
EINECS: 213-213-0
UN: 1224
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4830 (20°C, 589 nm)
GC ≥97.5 %
UV at 280 nm A: ≤0.04

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty