Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloropropene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11369-1000 11369-5000
TS11369-1000EA 126 USD InStock
TS11369-1000 TS11369-5000
2,3-Dichloropropene 98%
2,3-Dichloropropene


Formula: C₃H₄Cl₂
Boiling Pt: 94 °C (760 mmHg)
Melting Pt: –83.1 °C
Density: 1.2 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000943
CAS Number: 78-88-6
EINECS: 201-153-8
UN: 2047
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to pink-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4601 to 1.4621 (20°C, 589 nm)
GC ≥97.5 %
UV at 280 nm A: ≤0.04
Chloride (Cl) ≤500 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty