Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylbutane 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11610-0010 11610-0050 11610-1000
TS11610-0010EA 520 USD InStock
TS11610-0010 TS11610-0050 TS11610-1000
2,3-Dimethylbutane 98+%
2,3-Dimethylbutane


Formula: C₆H₁₄
Boiling Pt: 58 °C (760 mmHg)
Melting Pt: –129 °C
Density: 0.662 g/cm³
Flash Pt: –29 °C
Storage Temperature: Ambient
MDL Number: MFCD00008925
CAS Number: 79-29-8
EINECS: 201-193-6
UN: 2457
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3740 to 1.3760 (20°C, 589 nm)
GC ≥98.0 %
UV at 260 nm A: ≤0.01
Titration after Ion exchange ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty