Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl phosphite (contains varying amounts of phenol and (C₆H₅O)₃P)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11739-0010 11739-0050 11739-5000
TS11739-0010EA 121 USD InStock
TS11739-0010 TS11739-0050 TS11739-5000
Diphenyl phosphite (contains varying amounts of phenol and (C₆H₅O)₃P)
Diphenyl phosphite


Formula: C₁₂H₁₁O₃P
Boiling Pt: 218…219 °C (26 mmHg)
Melting Pt: 12 °C
Density: 1.217 g/cm³
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00044497
CAS Number: 4712-55-4
EINECS: 225-202-8
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5565 to 1.5585 (20°C, 589 nm)
Color scale ≤50 APHA
Specific gravity (25/15.5°C) 1.2 to 1.22
Acid value ≤15 mg KOH/g
Iron (Fe) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty