Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-2-cyclohexen-1-one 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

11788-0100 11788-0250 11788-1000
TS11788-0100EA 50 USD InStock
TS11788-0100 TS11788-0250 TS11788-1000
3-Ethoxy-2-cyclohexen-1-one 99%
3-Ethoxy-2-cyclohexen-1-one


Formula: C₈H₁₂O₂
Boiling Pt: 76…78 °C (1 mmHg)
Density: 1.04 g/cm³
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001580
CAS Number: 5323-87-5
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5020 to 1.5060 (20°C, 589 nm)
GC ≥98.5 %
Insoluble matter ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty