Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glutaryl dichloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11999-0250 11999-1000
TS11999-0250EA 67 USD InStock
TS11999-0250 TS11999-1000
Glutaryl dichloride 97%
Glutaryl dichloride


Formula: C₅H₆Cl₂O₂
MW: 169.0059 g/mol
Density: 1.324 g/cm³
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00000755
CAS Number: 2873-74-7
EINECS: 220-711-1
UN: 2922
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4760 (20°C, 589 nm)
GC ≥96.0 %
Titration RCOCl ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty