Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanidinium carbonate 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12022-0010 12022-0051 12022-2500
TS12022-0010EA 95 USD InStock
TS12022-0010 TS12022-0051 TS12022-2500
Guanidinium carbonate 99+%
Guanidinium carbonate


Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
Melting Pt: 267…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00013029
CAS Number: 593-85-1
EINECS: 209-813-7
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥97.5 %
pH 10.5 to 11.5 (1 % in water)
Specific optical rotation -9° to -14.5° (20°C, 589 nm) (c=4 in H2O)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty