Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1,3-cyclohexanedione 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12659-0250 12659-1000 12659-2000
TS12659-0250EA 128 USD InStock
TS12659-0250 TS12659-1000 TS12659-2000
2-Methyl-1,3-cyclohexanedione 98+%
2-Methyl-1,3-cyclohexanedione


Formula: C₇H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00001587
CAS Number: 1193-55-1
EINECS: 212-153-2
Order Now
Specification Test Results
Appearance Off-white to pink-orange to light brown Powder or crystals
Infrared spectrum Conforms
Melting point ≥204 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty