Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthyl isothiocyanate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12824-0050 12824-0100
TS12824-0050EA 94 USD InStock
TS12824-0050 TS12824-0100
1-Naphthyl isothiocyanate 98%
1-Naphthyl isothiocyanate


Formula: C₁₁H₇NS
Boiling Pt: 100 °C (0.2 mmHg)
Melting Pt: 55.5…57 °C
MDL Number: MFCD00003882
CAS Number: 551-06-4
EINECS: 208-990-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,06498
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥97.5 %
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 % (5 % in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty