Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propargyl amine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13146-0050 13146-0250 13146-1000
TS13146-0050EA 93 USD InStock
TS13146-0050 TS13146-0250 TS13146-1000
Propargyl amine 99%
Propargyl amine


Formula: C₃H₅N
Boiling Pt: 84 °C (760 mmHg)
Density: 0.86 g/cm³
Flash Pt: 3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008198
CAS Number: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4500 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty