Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propionyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13153-0025 13153-0250 13153-2500 13153-5000
TS13153-0025EA 192 USD InStock
TS13153-0025 TS13153-0250 TS13153-2500 TS13153-5000
Propionyl chloride 98%
Propionyl chloride


Formula: C₃H₅ClO
Boiling Pt: 77…79 °C (760 mmHg)
Melting Pt: –94 °C
Density: 1.06 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3,II
Merck Index: 12,08013
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4050 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤20 APHA
Titration RCOCl ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty