Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenylpropionyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13438-0250 13438-1000
TS13438-0250EA 87 USD InStock
TS13438-0250 TS13438-1000
3-Phenylpropionyl chloride 98%
3-Phenylpropionyl chloride


Formula: C₉H₉ClO
Boiling Pt: 107 °C (11 mmHg)
Melting Pt: –6 °C
Density: 1.137 g/cm³
Flash Pt: 108 °C
MDL Number: MFCD00000748
CAS Number: 645-45-4
EINECS: 211-443-6
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5245 to 1.5285 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty