Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Tropolone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

14058-0010 14058-0050 14058-0250
TS14058-0010EA 51 USD InStock
TS14058-0010 TS14058-0050 TS14058-0250
α-Tropolone 98%
α-Tropolone


Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 115 °C (12 mmHg)
Melting Pt: 50…54 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS Number: 533-75-5
EINECS: 208-577-2
Order Now
Specification Test Results
Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 54 °C (on dry substance)
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty