Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromopropene 80% tech.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

14774-0100 14774-0500 14774-2500
TS14774-0100EA 39 USD InStock
TS14774-0100 TS14774-0500 TS14774-2500
2,3-Dibromopropene 80% tech.
2,3-Dibromopropene


Formula: C₃H₄Br₂
Boiling Pt: 42…44 °C (17 mmHg)
Density: 2.13 g/cm³
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000211
CAS Number: 513-31-5
EINECS: 208-155-8
UN: 3334
ADR: 9,III
Merck Index: 13,03050
Order Now
Specification Test Results
Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
GC ≥75 %
Elemental Analysis 14.58 to 18.56 % (N)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty