Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyryl chloride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

14921-0010 14921-0500 14921-2500
TS14921-0010EA 158 USD InStock
TS14921-0010 TS14921-0500 TS14921-2500
Butyryl chloride 99%
Butyryl chloride


Formula: C₄H₇ClO
Boiling Pt: 102 °C (760 mmHg)
Melting Pt: –89 °C
Density: 1.018 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000752
CAS Number: 141-75-3
EINECS: 205-498-5
UN: 2353
ADR: 3,II
Merck Index: 13,01598
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4112 to 1.4132 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
Titration RCOOH ≤2 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty