Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloroaniline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15039-0010 15039-0250 15039-2500
TS15039-0010EA 248 USD InStock
TS15039-0010 TS15039-0250 TS15039-2500
2,4-Dichloroaniline 98%
2,4-Dichloroaniline


Formula: C₆H₅Cl₂N
Boiling Pt: 242 °C (760 mmHg)
Melting Pt: 59…63 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS Number: 554-00-7
EINECS: 209-057-8
UN: 3442
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Beige to light grey to light brown Powder or crystals or flakes and/or chunks (May darken on storage)
Infrared spectrum Conforms
Melting point 59 to 63 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty