Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminothiazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15389-1000 15389-5000
TS15389-1000EA 35 USD InStock
TS15389-1000 TS15389-5000
2-Aminothiazole 97%
2-Aminothiazole


Formula: C₃H₄N₂S
Boiling Pt: 117 °C (20mbar)
Melting Pt: 86…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00005325
CAS Number: 96-50-4
EINECS: 202-511-6
Merck Index: 13,00479
Order Now
Specification Test Results
Appearance Brown Powder or flakes or granules and/or lumps
Infrared spectrum Conforms
Melting point 86 to 91 °C
Water ≤0.75 % (K.F.)
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty