Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromocyclohexane 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15908-0010 15908-2500
TS15908-0010EA 161 USD InStock
TS15908-0010 TS15908-0025 TS15908-2500
Bromocyclohexane 99%
Bromocyclohexane


Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 166
Melting Pt: -56.5
Density: 1.33 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003819
CAS Number: 108-85-0
EINECS: 203-622-2
Merck Index: 13,02759
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty