Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromopropionyl chloride 95% tech.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16199-0250 16199-1000
TS16199-0250EA 83 USD InStock
TS16199-0250 TS16199-1000
3-Bromopropionyl chloride 95% tech.
3-Bromopropionyl chloride


Formula: BrH₂CCH₂COOCl
MW: 171.42 g/mol
Boiling Pt: 55
Density: 1.7 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000746
CAS Number: 15486-96-1
EINECS: 239-515-2
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4920 to 1.4970 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty